Learn More
Sino Biological Influenza A H14N5 (A/mallard/Astrakhan/263/1982) Hemagglutinin Protein (HA1 Subunit) Antibody, Mouse MAb
Supplier:  Sino Biological 40192MM0720
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A H14N5 (A/mallard/Astrakhan/263/1982) Hemagglutinin Protein (HA1 Subunit) ( Catalog#40192-V08H1; P26136; Met1-Lys342). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0704