Learn More
Labtech Inc H150-1500, 230V, 50/60Hz water chiller has a touchscreen display, temperature setting at 8-35°C, cooling capacity of 1500W at 25C degree. pump capacity of 6.4L/min and a max pressure of 60psi for external circulation.
Catalog No. 5000496199
Encompass_Preferred
H150-1500, 230V, 50/60Hz water chiller has a touchscreen display, temperature setting at 8-35°C, cooling capacity of 1500W at 25C degree. pump capacity of 6.4L/min and a max pressure of 60psi for external circulation.
Catalog No. 50-004-96199 Supplier Labtech Inc Supplier No. LW1500230
H150-1500, 230V, 50/60Hz water chiller has a touchscreen display, temperature setting at 8-35°C, cooling capacity of 1500W at 25C degree. pump capacity of 6.4L/min and a max pressure of 60psi for external circulation.