Learn More
Labtech Inc H150-2100, 230V, 50/60Hz water chiller has a touchscreen display, temperature setting at 8-35°C, cooling capacity of 2100W at 25C degree. pump capacity of 12.1L/min and a max pressure of 60psi for external circulation.
Catalog No. 5000496184
Encompass_Preferred
H150-2100, 230V, 50/60Hz water chiller has a touchscreen display, temperature setting at 8-35°C, cooling capacity of 2100W at 25C degree. pump capacity of 12.1L/min and a max pressure of 60psi for external circulation.
Catalog No. 50-004-96184 Supplier Labtech Inc Supplier No. LW2100
Add to Cart
Add to Cart
H150-2100, 230V, 50/60Hz water chiller has a touchscreen display, temperature setting at 8-35°C, cooling capacity of 2100W at 25C degree. pump capacity of 12.1L/min and a max pressure of 60psi for external circulation.