Learn More
Sino Biological Influenza A H15N8 (A/duck/AUS/341/1983) Hemagglutinin / HA Antibody, Rabbit PAb
Supplier:  Sino Biological 11720RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H15N8 (A/duck/AUS/341/1983) Hemagglutinin / HA (Catalog#11720-V08H; ABB88132.1; Met1-Val534). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0799