Learn More
Sino Biological Influenza A H15N8 (A/duck/AUS/341/1983) Hemagglutinin / HA Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 11720T6220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H15N8 (A/duck/AUS/341/1983) Hemagglutinin / HA (Catalog#11720-V08H; ABB88132.1; Met1-Val534). Influenza A H15N8 (A/duck/AUS/341/1983) Hemagglutinin / HA specific IgG was purified by Influenza A H15N8 (A/duck/AUS/341/1983) Hemagglutinin / HA affinity chromatography.
Catalog No. 50-240-8530