Learn More
Sino Biological Influenza A H16N3 Hemagglutinin / HA Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 11711RP0220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant H16N3 (A/black-headed gull/Sweden/5/99) HA / Hemagglutinin (Catalog#11711-V08H; AAV91217.1; Met 1-Lys 529). H16N3 (A/black-headed gull/Sweden/5/99) HA / Hemagglutinin specific IgG was purified by H16N3 (A/black-headed gull/Sweden/5/99) HA / Hemagglutinin affinity chromatography.
Catalog No. 50-240-9316