Learn More
Sino Biological Influenza A H17N10 (A/little yellow-shouldered bat/Guatemala/164/2009) Hemagglutinin Antibody, Mouse MAb
Supplier:  Sino Biological 40323MM0920
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A H17N10 (A/little yellow-shouldered bat/Guatemala/164/2009) Hemagglutinin ( Catalog#40323-V08H1; AFC35418.1; Met1-Tyr527). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-3723