Learn More
Sino Biological Influenza A H17N10 (A/little yellow-shouldered bat/Guatemala/164/2009) Hemagglutinin Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 40323T6220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H17N10 (A/little yellow-shouldered bat/Guatemala/164/2009) Hemagglutinin ( Catalog#40323-V08H1; AFC35418.1; Met1-Tyr527). Influenza A H17N10 (A/little yellow-shouldered bat/Guatemala/164/2009) Hemagglutinin specific IgG was purified by Influenza A H17N10 (A/little yellow-shouldered bat/Guatemala/164/2009) Hemagglutinin affinity chromatography.
Catalog No. 50-241-1206