Learn More
Sino Biological Influenza A H18N11 (A/flat-faced bat/Peru/033/2010) Hemagglutinin / HA1 Antibody, Mouse MAb
Supplier:  Sino Biological 40324MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A H18N11 (A/flat-faced bat/Peru/033/2010) Hemagglutinin / HA1 ( Catalog#40324-V08H1; AGX84934.1; Met1-Arg339). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8678