Learn More
Aviva Systems Biology Corp H1FOO Antibody - middle region (ARP53205_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53205_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against H1FOO. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human H1FOO. Alternate names: MGC50807, osH1ApplicationWB. Species reactivity: Human
Catalog No. 50-285-808