Learn More
Biotang Inc Influenza A H1N1 (A/California/04/2009) Hemagglutinin / HA Protein (His Tag), 10ug, extracellular domain of Influenza A virus hemagglutinin (ACP41105.1) (Met 1-Gln 529) with cleavage site mutated (IQS-RQRRRRK, HA1+HA2, partially cleaved)
Supplier:  Biotang Inc RPR9503
Encompass_Preferred
Influenza A H1N1 (A/California/04/2009) Hemagglutinin / HA Protein (His Tag), 10ug, extracellular domain of Influenza A virus hemagglutinin (ACP41105.1) (Met 1-Gln 529) with cleavage site mutated (IQS-RQRRRRK, HA1+HA2, partially cleaved)
Catalog No. 50-751-5058