Learn More
Creative Biomart Recombinant CoV-2 Membrane Env Antigen, His-tagged
Supplier:  Creative Biomart H1V1302
Encompass_Preferred
The E. coli derived recombinant fusion protein contains the Coronavirus 2019 Full-Length Membrane and Envelope proteins, Wuhan-Hu-1 strain, having a Mw of 34.2 kDa fused to His tag at C-terminal.
Catalog No. 50-208-4680