Learn More
Sino Biological Influenza A H1N1 (A/Victoria/4897/2022) Hemagglutinin / HA-specific B cell probe Protein (Y108F, AVI & His Tag), Biotinylated
Catalog No. 502993255
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Victoria/4897/2022 (H1N1)) hemagglutinin (EPI2805564) (Met1-Asp519, with a mutation Tyr108Phe), termed as HA, was expressed with a c-terminal AVI tagged polyhistidine tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-3255 Supplier Sino Biological Supplier No. 41049V27HB20
A DNA sequence encoding the Influenza A virus (A/Victoria/4897/2022 (H1N1)) hemagglutinin (EPI2805564) (Met1-Asp519, with a mutation Tyr108Phe), termed as HA, was expressed with a c-terminal AVI tagged polyhistidine tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.