Learn More
Sino Biological Influenza A H1N1 (strain A/Puerto Rico/8/1934) Hemagglutinin / HA-specific B cell probe Protein (Y108F, AVI & His Tag), Biotinylated
Catalog No. 502993516
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (strain A/Puerto Rico/8/1934 (H1N1)) hemagglutinin (P03452) (Met1-Gly518, with a mutation Tyr108Phe), termed as HA, was expressed with a c-terminal AVI tagged polyhistidine tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-3516 Supplier Sino Biological Supplier No. 11684V27HB20
A DNA sequence encoding the Influenza A virus (strain A/Puerto Rico/8/1934 (H1N1)) hemagglutinin (P03452) (Met1-Gly518, with a mutation Tyr108Phe), termed as HA, was expressed with a c-terminal AVI tagged polyhistidine tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.