Learn More
Sino Biological Influenza A H1N1 (A/California/07/09) Hemagglutinin / HA trimer Protein (His & AVI Tag), Biotinylated
Catalog No. 502992848
Encompass_Preferred
A DNA sequence encoding the Influenza A virus ((A/California/07/09) (H1N1)) hemagglutinin (translated amino acids of ACP41953.1), termed as HA, was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-2848 Supplier Sino Biological Supplier No. 11085V49H1B20
A DNA sequence encoding the Influenza A virus ((A/California/07/09) (H1N1)) hemagglutinin (translated amino acids of ACP41953.1), termed as HA, was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.