Learn More
Sino Biological Influenza A H1N1 (Swine Flu 2009) Hemagglutinin / HA Antibody, Mouse MAb
Supplier:  Sino Biological 11055MM08100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A virus H1N1 hemagglutinin (HA) extracellular domain. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4925