Learn More
Sino Biological H1N1 HA MAb 20ug (Sino Biological)
Supplier:  Sino Biological 11055MM1120
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A virus H1N1 hemagglutinin (HA) extracellular domain. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2273