Learn More
Sino Biological Influenza A H1N1 (Swine Flu 2009) Hemagglutinin / HA Antibody (HRP), Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 11055RP07100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A virus H1N1 (Swine Flu 2009) Hemagglutinin extracellular domain. H1N1 Hemagglutinin / HA specific IgG was purified by Influenza A virus H1N1 Hemagglutinin / HA affinity chromatography, and conjugated with horseradish peroxidase (HRP).
Catalog No. 50-255-4918