Learn More
Sino Biological Influenza A H1N1 (A/New Caledonia/20/1999) Hemagglutinin / HA Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 11683T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H1N1 (A/New Caledonia/20/1999) Hemagglutinin / HA ( Catalog#11683-V08H; AAP34324.1; Met1-Gln528). Influenza A H1N1 (A/New Caledonia/20/1999) Hemagglutinin / HA specific IgG was purified by Influenza A H1N1 (A/New Caledonia/20/1999) Hemagglutinin / HA affinity chromatography.
Catalog No. 50-255-4615