Learn More
Sino Biological Influenza A H1N1 (A/California/04/2009) Hemagglutinin / HA Antibody (Biotin), Rabbit PAb
Supplier:  Sino Biological 11055RP01B20
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H1N1 (A/California/04/2009) Hemagglutinin / HA. Total IgG was purified by Protein A affinity chromatography and then biotinylated.
Catalog No. 50-255-4920