Learn More
Sino Biological Influenza A H1N1 (A/California/04/2009) Hemagglutinin / HA Antibody (HRP), Rabbit PAb
Supplier:  Sino Biological 11055RP01H20
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H1N1 (A/California/04/2009) Hemagglutinin / HA. Total IgG was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-255-4919