Learn More
Sino Biological H1N1 HA PAb 20ul (Sino Biological)
Supplier:  Sino Biological 11684RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H1N1 (A/Puerto Rico/8/1934) hemagglutinin (Catalog#11684-V08H; ABD77675.1; Met 1-Gln 528). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-2014