Learn More
Sino Biological H1N1 HA PAb 20ul (Sino Biological)
Supplier:  Sino Biological 11068RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H1N1 (A/Brevig Mission/1/1918) Hemagglutinin / HA (Catalog#11068-V08H; AAD17229.1; Met1-Gln529). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-2246