Learn More
Sino Biological H1N1 HA PAb 20ul (Sino Biological)
Supplier:  Sino Biological 11055T6220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H1N1 (A/California/04/2009) Hemagglutinin / HA ( Catalog#11055-V08H; ACP41105.1; Met1-Gln529). Influenza A H1N1 (A/California/04/2009) Hemagglutinin / HA specific IgG was purified by Influenza A H1N1 (A/California/04/2009) Hemagglutinin / HA affinity chromatography.
Catalog No. 50-237-3073