Learn More
Sino Biological Influenza A H1N1 (A/swine/Henan/SN13/2018) Hemagglutinin / HA Protein (ECD, His Tag)
Supplier:  Sino Biological 40648V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A H1N1 (A/swine/Henan/SN13/2018) Hemagglutinin / HA (QEM24688.1) (Met1-Ile530) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3689