Learn More
Sino Biological Influenza A H1N1 (A/swine/Hebei/0116/2017) Hemagglutinin / HA Protein (ECD, His Tag)
Supplier:  Sino Biological 40645V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/swine/Hebei/0116/2017 (H1N1)) hemagglutinin (QEM24663.1) (Met1-Ile530) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3691