Learn More
Sino Biological Influenza A H1N2 (A/swine/Kansas/00246/2004) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40665V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/swine/Kansas/00246/2004 (H1N2)) hemagglutinin (ABV25642)(Met1-Ser524), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3671