Learn More
Sino Biological Influenza A H1N1 (A/Puerto Rico/8/1934) Hemagglutinin / HA trimer Protein (His Tag)(MALS-verified)
Supplier:  Sino Biological 11684V08H2100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus ((A/Puerto Rico/8/1934) (H1N1)) hemagglutinin termed as HA, was expressed with the bacteriophage T4 fibritin and a polyhistidine tag at the C-terminus.
Catalog No. 50-241-2430