Learn More
Sino Biological Influenza A H1N1 (A/swine/England/267/2007) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40702V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/swine/England/267/2007 (H1N1)) hemagglutinin (AKJ81829) (Met1-Ser524), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3637