Learn More
Sino Biological Influenza A H1N1 (A/swine/Mexico/AVX47/2013) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40681V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/swine/Mexico/AVX47/2013 (H1N1)) hemagglutinin (AMY16048) (Ala17-Ser524), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3656