Learn More
Sino Biological Influenza A H1N1 (A/Wisconsin/67/2022) Hemagglutinin / HA Protein (ECD, His Tag)
Supplier:  Sino Biological 40940V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus ((A/Wisconsin/67/2022) (H1N1)) hemagglutinin (translated amino acids of EPI2224788) (Met1-Ile530), termed as HA, was expressed with the bacteriophage T4 fibritin and a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3741