Learn More
Sino Biological H1N1 HA Protein 100ug (Sino Biological)
Supplier:  Sino Biological 40940V08H2100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus ((A/Wisconsin/67/2022) (H1N1)) hemagglutinin (translated amino acids of EPI2224788), termed as HA, was expressed with the bacteriophage T4 fibritin and a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3898