Learn More
Sino Biological Influenza A H1N1 (A/Puerto Rico/8/34/Mount Sinai) Matrix protein 1 / M1 Antibody, Rabbit PAb
Supplier:  Sino Biological 40010RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H1N1 (A/Puerto Rico/8/34/Mount Sinai) Matrix protein 1 / M1 (Catalog#40010-V07E; NP_040978.1; Ser2-Lys252). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1966