Learn More
Sino Biological Influenza A H1N1 (A/California/04/2009) Neuraminidase / NA Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 11058T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H1N1 (A/California/04/2009) Neuraminidase / NA ( Catalog#11058-V01H; ACP41107.1; His36-Lys469). Neuraminidase/NA specific IgG was purified by Influenza A H1N1 (A/California/04/2009) Neuraminidase / NA affinity chromatography.
Catalog No. 50-237-4222