Learn More
Sino Biological H1N1 NA Protein 100ug (Sino Biological)
Supplier:  Sino Biological 40939V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Victoria/4897/2022 (H1N1)) Neuraminidase / NA (EPI2397518) (Ile30-Asn469) was expressed with a vasodilator-stimulated phosphoprotein tetramerization domain at the N-terminus and a polyhistidine tag at the C-terminus.
Catalog No. 50-237-1770