Learn More
Sino Biological Influenza A H1N1 (A/Auckland/6/1996) Neuraminidase / NA (His Tag)
Supplier:  Sino Biological 40731V07H100
Encompass_Preferred
A DNA sequence encoding the influenza A virus (A/Auckland/6/1996 (H1N1)) neuraminidase (ABQ53689) (His36-Lys470), termed as NA, was fused with a N-terminal polyhistidine tag.
Catalog No. 50-255-3616