Learn More
Sino Biological Influenza A H1N1 (A/AA/Marton/1943) Neuraminidase / NA (His Tag)
Supplier:  Sino Biological 40727V07H100
Encompass_Preferred
A DNA sequence encoding the influenza A virus (A/AA/Marton/1943 (H1N1)) neuraminidase (ABO38057) (His36-Lys469), termed as NA, was fused with a N-terminal polyhistidine tag.
Catalog No. 50-255-3620