Learn More
Sino Biological Influenza A H1N1 (A/swine/Shandong/1207/2016) Neuraminidase / NA (His Tag)
Supplier:  Sino Biological 40652V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/swine/Shandong/1207/2016 (H1N1)) neuraminidase (QEM24782.1) (Ile30-Lys469) was expressed with a vasodilator-stimulated phosphoprotein tetramerization domain at the N-terminus and a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3684