Learn More
Sino Biological Influenza A H1N1 (A/Sw/Bulnes/VN1401-P6SP/2018) Neuraminidase / NA (His Tag)
Supplier:  Sino Biological 40738V07H100
Encompass_Preferred
A DNA sequence encoding the influenza A virus (A/Sw/Bulnes/VN1401-P6SP/2018 (H1N1)) neuraminidase (AYV62750) (His36-Lys469), termed as NA, was fused with a N-terminal polyhistidine tag.
Catalog No. 50-255-3610