Learn More
Sino Biological Influenza A H1N1 (A/Sydney/5/2021) Neuraminidase / NA Protein (ECD, His Tag)
Supplier:  Sino Biological 40952V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Sydney/5/2021 (H1N1)) Neuraminidase / NA (EPI2413554) (Ile30-Lys469) was expressed with a vasodilator-stimulated phosphoprotein tetramerization domain at the N-terminus and a polyhistidine tag at the C-terminus.
Catalog No. 50-240-8413