Learn More
Sino Biological Influenza A H1N1 (A/Wisconsin/67/2022) Neuraminidase / NA Protein (ECD, His Tag)
Supplier:  Sino Biological 40941V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Wisconsin/67/2022 (H1N1)) Neuraminidase / NA (EPI2224787) (Ile30-Asn469) was expressed with a vasodilator-stimulated phosphoprotein tetramerization domain at the N-terminus and a polyhistidine tag at the C-terminus.
Catalog No. 50-237-1160