Learn More
Creative Biomart Recombinant Dengue Premembrane Antigen, His-tagged
Catalog No. 502084682
Encompass_Preferred
Recombinant dengue 2 premembrane is a whole length peptide expressed from E. coil with 18 kDa in size. PreM is considered as a signal peptide responsible for dengue envelope translocation in cells. Ex
Catalog No. 50-208-4682 Supplier Creative Biomart Supplier No. H1V1304
 Restricted Item
Add to Cart
Add to Cart
Recombinant dengue 2 premembrane is a whole length peptide expressed from E. coil with 18 kDa in size. PreM is considered as a signal peptide responsible for dengue envelope translocation in cells. Ex