Learn More
Sino Biological H1N1 NP ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502372619
Encompass_Preferred
Each vial contains 56 ng of recombinant Influenza A H1N1 (A/California/07/2009) Nucleoprotein. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 4000 pg/mL is recommended.
Catalog No. 50-237-2619 Supplier Sino Biological Supplier No. SEK00415
Each vial contains 56 ng of recombinant Influenza A H1N1 (A/California/07/2009) Nucleoprotein. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 4000 pg/mL is recommended.