Learn More
Sino Biological Influenza A H1N1 (A/Brisbane/02/2018) Nucleoprotein / NP Protein (His Tag)
Supplier:  Sino Biological 40776V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Brisbane/02/2018 (H1N1)) nucleoprotein (translated amino acids of EPI1383382) (Met1-Asn498) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3591