Learn More
Sino Biological Influenza A H1N1 (A/Wisconsin/67/2022) Nucleoprotein/NP (His Tag)
Supplier:  Sino Biological 40942V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus ((A/Wisconsin/67/2022) (H1N1)) nucleoprotein (EPI2224781) (Met1-Asn498) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-240-7848