Learn More
Sino Biological Influenza A H1N1 (A/Puerto Rico/8/34/Mount Sinai) Non-structural Protein 2 / NS2
Supplier:  Sino Biological 40012VNAE100
Encompass_Preferred
A DNA sequence encoding the influenza A H1N1 Virus (A/Puerto Rico/8/34/Mount Sinai) NS2 protein (P03508.1) (Met 1-Ile 121) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-3767