Learn More
Creative Biomart Recombinant HBsAg ayw Antigen, His-tagged
Supplier:  Creative Biomart H1V1306
Encompass_Preferred
Recombinant HbsAg ayw full length is a 31 kDa proteincloned from HBV 320 genome and fused to 6 His tag.
Catalog No. 50-208-4684