Learn More
Sino Biological H1N2 HA PAb 20ul (Sino Biological)
Supplier:  Sino Biological 11703RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant H1N2 (A/swine/Guangxi/13/2006) HA / Hemagglutinin (Catalog#11703-V08H; ABQ42444.1; Met 1-Gln 529). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-2532