Learn More
Sino Biological Influenza A H1N2 (A/Minnesota/14/2012) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40677V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Minnesota/14/2012 (H1N2)) hemagglutinin (AHW83238) (Met1-Ser523), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3660