Learn More
Sino Biological Influenza A H1N2 (A/swine/North Carolina/A01668056/2016) Neuraminidase / NA (His Tag)
Supplier:  Sino Biological 40749V07H100
Encompass_Preferred
A DNA sequence encoding the influenza A virus (A/swine/North Carolina/A01668056/2016 (H1N2)) neuraminidase (APQ31966) (His36-Val469), termed as NA, was fused with a N-terminal polyhistidine tag.
Catalog No. 50-255-3599